Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300426.1 | NP_001287355.1 | 6 - 7 | 2299 | 112 | |
NM_057361.4 | NP_476709.1 | 6 - 7 | 2055 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |