Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170304.3 | NP_001163775.1 | 5 - 6 | 1542 | 92 | |
NM_001170305.3 | NP_001163776.1 | 6 - 7 | 1825 | 92 | ||
NM_001170306.2 | NP_001163777.1 | 6 - 7 | 1578 | 92 | ||
NM_001202391.2 | NP_001189320.1 | 6 - 7 | 1487 | 92 | ||
NM_057683.5 | NP_477031.1 | 5 - 6 | 1362 | 92 | ||
NM_057684.4 | NP_477032.1 | 5 - 6 | 1333 | 92 | ||
NM_170503.4 | NP_733382.1 | 5 - 6 | 1731 | 92 | ||
NM_170504.4 | NP_733383.1 | 6 - 7 | 1510 | 92 | ||
NM_170505.4 | NP_733384.1 | 6 - 7 | 1595 | 92 | ||
NM_170506.3 | NP_733385.1 | 6 - 7 | 1541 | 92 | ||
GenBank mRNA | AY094727.1 | - | 5 - 6 | 1333 | 92 | |
BT133205.1 | - | 5 - 6 | 1334 | 92 | ||
U92490.1 | - | 5 - 6 | 1371 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |