Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ask1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057741.4 | NP_477089.3 | 5 - 6 | 1932 | 78 | |
NM_206518.2 | NP_996240.1 | 4 - 5 | 2504 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |