Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sodh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300274.1 | NP_001287203.1 | 3 - 4 | 628 | 87 | |
NM_058000.3 | NP_477348.1 | 3 - 4 | 820 | 87 | ||
GenBank mRNA | BT044539.1 | - | 3 - 4 | 820 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |