Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058116.5 | NP_477464.1 | 3 - 4 | 1414 | 67 | |
GenBank mRNA | AF181098.1 | - | 3 - 4 | 1376 | 67 | |
BT133170.1 | - | 3 - 4 | 1418 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |