Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Est9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079534.3 | NP_524258.2 | 2 - 3 | 905 | 64 | |
NM_169187.3 | NP_731165.2 | 2 - 3 | 905 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |