Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or85b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079555.2 | NP_524279.2 | 1 - 2 | 967 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Drosophila melanogaster |