Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275474.1 | NP_001262403.1 | 4 - 5 | 2383 | 86 | |
NM_079561.4 | NP_524285.2 | 4 - 5 | 2383 | 86 | ||
NM_169258.3 | NP_731314.1 | 4 - 5 | 2235 | 86 | ||
NM_169259.3 | NP_731315.1 | 4 - 5 | 2233 | 86 | ||
NM_169260.4 | NP_731316.2 | 2 - 3 | 868 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |