Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275482.1 | NP_001262411.1 | 9 - 10 | 2187 | 66 | |
NM_001275483.1 | NP_001262412.1 | 9 - 10 | 2190 | 66 | ||
NM_001382200.1 | NP_001368998.1 | 2 - 3 | 678 | 66 | ||
NM_079564.4 | NP_524288.3 | 9 - 10 | 3549 | 66 | ||
NM_169274.2 | NP_731341.1 | 8 - 9 | 1986 | 66 | ||
NM_169275.2 | NP_731342.1 | 6 - 7 | 1427 | 66 | ||
NM_169276.2 | NP_731343.1 | 2 - 3 | 894 | 66 | ||
GenBank mRNA | BT003462.1 | - | 9 - 10 | 3547 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |