Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nerfin-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275517.1 | NP_001262446.1 | 1 - 2 | 1529 | 64 | |
NM_079576.3 | NP_524300.1 | 1 - 2 | 1529 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |