Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260137.1 | NP_001247066.1 | 4 - 5 | 615 | 67 | |
NM_079600.3 | NP_524324.2 | 5 - 6 | 945 | 67 | ||
NM_169444.1 | NP_731661.1 | 5 - 6 | 633 | 67 | ||
NM_176466.1 | NP_788643.1 | 5 - 6 | 652 | 67 | ||
NM_176467.1 | NP_788644.1 | 5 - 6 | 630 | 67 | ||
NM_176468.1 | NP_788645.1 | 4 - 5 | 1029 | 67 | ||
NM_176469.2 | NP_788646.1 | 5 - 6 | 750 | 67 | ||
NM_176470.1 | NP_788647.1 | 5 - 6 | 655 | 67 | ||
NM_176471.1 | NP_788648.1 | 5 - 6 | 828 | 67 | ||
NM_176472.1 | NP_788649.1 | 5 - 6 | 850 | 67 | ||
NM_176473.1 | NP_788650.1 | 6 - 7 | 724 | 67 | ||
NM_176474.1 | NP_788651.1 | 6 - 7 | 702 | 67 | ||
NM_176475.1 | NP_788652.1 | 5 - 6 | 1101 | 67 | ||
NM_176476.1 | NP_788653.1 | 6 - 7 | 705 | 67 | ||
NM_176477.1 | NP_788654.1 | 6 - 7 | 727 | 67 | ||
NM_176478.1 | NP_788655.1 | 6 - 7 | 900 | 67 | ||
GenBank mRNA | AY069094.1 | - | 5 - 6 | 615 | 67 | |
BT120190.1 | - | 4 - 5 | 615 | 67 | ||
BT125878.1 | - | 6 - 7 | 701 | 67 | ||
BT133296.1 | - | 6 - 7 | 879 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |