Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Past1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079608.3 | NP_524332.2 | 1 - 2 | 1505 | 81 | |
NM_169478.2 | NP_731737.1 | 2 - 3 | 1434 | 81 | ||
GenBank mRNA | AF473822.1 | - | 1 - 2 | 1225 | 81 | |
AY075248.1 | - | 2 - 3 | 1434 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |