Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Past1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079608.3 | NP_524332.2 | 2 - 3 | 1677 | 64 | |
NM_169478.2 | NP_731737.1 | 3 - 4 | 1606 | 64 | ||
GenBank mRNA | AF473822.1 | - | 2 - 3 | 1397 | 64 | |
AY075248.1 | - | 3 - 4 | 1606 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |