Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cher | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202309.1 | NP_001189238.1 | 8 - 9 | 3706 | 63 | |
NM_001275728.1 | NP_001262657.2 | 10 - 11 | 4583 | 63 | ||
NM_001275729.1 | NP_001262658.1 | 9 - 10 | 4273 | 63 | ||
NM_001275730.1 | NP_001262659.2 | 11 - 12 | 4598 | 63 | ||
NM_079659.4 | NP_524383.3 | 8 - 9 | 3767 | 63 | ||
NM_206502.3 | NP_996224.2 | 9 - 10 | 4334 | 63 | ||
GenBank mRNA | AF188360.1 | - | 8 - 9 | 3743 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |