Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079664.2 | NP_524388.1 | 1 - 2 | 303 | 75 | |
GenBank mRNA | AY118986.1 | - | 1 - 2 | 275 | 75 | |
KX531913.1 | - | 1 - 2 | 273 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |