Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275793.1 | NP_001262722.1 | 3 - 4 | 818 | 76 | |
NM_079675.3 | NP_524399.1 | 2 - 3 | 746 | 76 | ||
GenBank mRNA | AF045610.2 | - | 2 - 3 | 722 | 76 | |
AY102689.1 | - | 2 - 3 | 760 | 76 | ||
Y18048.1 | - | 598 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |