Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260262.2 | NP_001247191.1 | 4 - 5 | 1794 | 63 | |
NM_079677.4 | NP_524401.2 | 4 - 5 | 1794 | 63 | ||
GenBank mRNA | BT004492.1 | - | 3 - 4 | 840 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |