Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170195.3 | NP_001163666.1 | 4 - 5 | 674 | 65 | |
NM_079696.6 | NP_524420.1 | 4 - 5 | 674 | 65 | ||
NM_169916.3 | NP_732544.1 | 4 - 5 | 674 | 65 | ||
GenBank mRNA | AY051671.1 | - | 4 - 5 | 633 | 65 | |
X57486.1 | - | 677 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |