Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170210.2 | NP_001163681.1 | 7 - 8 | 1491 | 60 | |
NM_001275882.1 | NP_001262811.1 | 7 - 8 | 1491 | 60 | ||
NM_079717.5 | NP_524441.2 | 7 - 8 | 1491 | 60 | ||
GenBank mRNA | BT046172.1 | - | 7 - 8 | 1475 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |