Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275316.2 | NP_001262245.1 | 2 - 3 | 718 | 56 | |
NM_001300226.1 | NP_001287155.1 | 3 - 4 | 1870 | 56 | ||
NM_141187.4 | NP_649444.1 | 3 - 4 | 1325 | 56 | ||
GenBank mRNA | AY075518.1 | - | 3 - 4 | 1325 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |