Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA1-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144539.2 | NP_001138011.1 | 10 - 11 | 3060 | 76 | |
NM_001275364.2 | NP_001262293.1 | 12 - 13 | 2715 | 76 | ||
NM_141309.4 | NP_649566.2 | 11 - 12 | 2607 | 76 | ||
NM_169086.3 | NP_730976.1 | 12 - 13 | 2715 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |