Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141371.3 | NP_649628.2 | 2 - 3 | 592 | 76 | |
GenBank mRNA | BT030794.1 | - | 2 - 3 | 587 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |