Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141477.2 | NP_649734.2 | 4 - 5 | 1165 | 74 | |
NM_169192.3 | NP_731174.1 | 3 - 4 | 732 | 74 | ||
NM_169193.3 | NP_731175.2 | 3 - 4 | 732 | 74 | ||
GenBank mRNA | AF233093.1 | - | 3 - 4 | 723 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |