Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmdyn-D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141669.3 | NP_649926.2 | 3 - 4 | 600 | 66 | |
GenBank mRNA | BT150178.1 | - | 3 - 4 | 598 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |