Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141674.3 | NP_649931.1 | 5 - 6 | 2238 | 70 | |
GenBank mRNA | AY058438.1 | - | 5 - 6 | 2200 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |