Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpt3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141681.4 | NP_649938.2 | 1 | 179 | 105 | |
NM_169308.4 | NP_731401.3 | 2 | 161 | 105 | ||
GenBank mRNA | BT016035.1 | - | 1 | 165 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |