Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eloF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141699.3 | NP_649956.1 | 1 - 2 | 222 | 100 | |
GenBank mRNA | BT024993.1 | - | 1 - 2 | 222 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |