Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Teh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275513.1 | NP_001262442.1 | 2 - 3 | 1824 | 85 | |
NM_141702.3 | NP_649959.1 | 2 - 3 | 1824 | 85 | ||
GenBank mRNA | AY059448.1 | - | 2 - 3 | 1823 | 85 | |
DQ139957.1 | - | 1 - 2 | 770 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |