Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141863.3 | NP_650120.1 | 6 - 7 | 1641 | 68 | |
NM_169434.2 | NP_731639.1 | 6 - 7 | 1566 | 68 | ||
NM_169435.3 | NP_731640.1 | 5 - 6 | 1496 | 68 | ||
GenBank mRNA | AY089465.1 | - | 6 - 7 | 1723 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |