Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300362.1 | NP_001287291.1 | 1 - 2 | 589 | 73 | |
NM_141869.3 | NP_650126.2 | 1 - 2 | 589 | 73 | ||
GenBank mRNA | BT100276.1 | - | 1 - 2 | 586 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Drosophila melanogaster |