Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-Vc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001369977.1 | NP_001356887.1 | 6 - 7 | 1611 | 79 | |
GenBank mRNA | BT088886.1 | - | 5 - 6 | 1188 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |