Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Task6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275610.1 | NP_001262539.1 | 6 - 7 | 1903 | 112 | |
NM_001275611.1 | NP_001262540.1 | 6 - 7 | 2951 | 112 | ||
NM_001275612.1 | NP_001262541.1 | 6 - 7 | 2951 | 112 | ||
NM_142043.3 | NP_650300.2 | 6 - 7 | 1903 | 112 | ||
GenBank mRNA | AJ920330.1 | - | 5 - 6 | 791 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |