Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Task6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275612.1 | NP_001262541.1 | 8 - 9 | 3140 | 81 | |
NM_142043.3 | NP_650300.2 | 8 - 9 | 2092 | 81 | ||
GenBank mRNA | AJ920330.1 | - | 7 - 8 | 980 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster Inventoried |