Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142050.3 | NP_650307.2 | 1 - 2 | 183 | 61 | |
GenBank mRNA | AJ276682.1 | - | 1 - 2 | 47 | 61 | |
AY121617.1 | - | 1 - 2 | 158 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |