Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR-C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142144.3 | NP_650401.1 | 3 - 4 | 334 | 69 | |
NM_169607.2 | NP_731954.1 | 2 - 3 | 406 | 69 | ||
GenBank mRNA | AY058243.1 | - | 2 - 3 | 287 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |