Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300410.1 | NP_001287339.1 | 2 - 3 | 425 | 66 | |
NM_142209.3 | NP_650466.2 | 2 - 3 | 489 | 66 | ||
NM_169657.3 | NP_732033.1 | 2 - 3 | 1172 | 66 | ||
NM_169658.3 | NP_732034.2 | 2 - 3 | 489 | 66 | ||
GenBank mRNA | AY095516.1 | - | 2 - 3 | 924 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |