Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275724.1 | NP_001262653.1 | 9 - 10 | 2098 | 100 | |
NM_142327.3 | NP_650584.1 | 9 - 10 | 2098 | 100 | ||
GenBank mRNA | AY051443.1 | - | 9 - 10 | 2076 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |