Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275748.1 | NP_001262677.1 | 1 - 2 | 301 | 67 | |
NM_142387.3 | NP_650644.1 | 1 - 2 | 301 | 67 | ||
GenBank mRNA | BT010086.1 | - | 1 - 2 | 272 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |