Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TyrRII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275753.1 | NP_001262682.1 | 3 - 4 | 661 | 58 | |
NM_001275754.2 | NP_001262683.1 | 3 - 4 | 1299 | 58 | ||
NM_001300451.1 | NP_001287380.1 | 3 - 4 | 1299 | 58 | ||
NM_142394.2 | NP_650651.1 | 2 - 3 | 862 | 58 | ||
GenBank mRNA | BT029098.1 | - | 2 - 3 | 725 | 58 | |
DQ151546.1 | - | 2 - 3 | 862 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |