Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142667.3 | NP_650924.3 | 7 - 8 | 1171 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |