Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275874.1 | NP_001262803.1 | 3 - 4 | 498 | 75 | |
NM_142696.3 | NP_650953.1 | 3 - 4 | 498 | 75 | ||
GenBank mRNA | BT024286.1 | - | 3 - 4 | 498 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |