Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pinta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300537.1 | NP_001287466.1 | 2 - 3 | 857 | 63 | |
NM_142785.2 | NP_651042.1 | 2 - 3 | 653 | 63 | ||
GenBank mRNA | BT005203.1 | - | 2 - 3 | 620 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |