Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142836.3 | NP_651093.1 | 1 | 370 | 90 | |
GenBank mRNA | AY061322.1 | - | 1 | 329 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |