Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142879.3 | NP_651136.1 | 1 - 2 | 254 | 111 | |
GenBank mRNA | AY118904.1 | - | 1 - 2 | 223 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |