Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir94g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142890.2 | NP_651147.2 | 1 | 1590 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |