Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SdhD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142924.3 | NP_651181.1 | 2 - 3 | 267 | 97 | |
GenBank mRNA | AY071388.1 | - | 2 - 3 | 264 | 97 | |
AY071617.1 | - | 2 - 3 | 264 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |