Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275966.1 | NP_001262895.1 | 2 - 3 | 1855 | 67 | |
NM_142948.4 | NP_651205.2 | 1 - 2 | 800 | 67 | ||
NM_170111.3 | NP_732936.1 | 1 - 2 | 800 | 67 | ||
NM_170114.3 | NP_732939.2 | 1 - 2 | 800 | 67 | ||
GenBank mRNA | BT016011.1 | - | 2 - 3 | 1855 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |