Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275967.2 | NP_001262896.1 | 4 - 5 | 1639 | 92 | |
NM_001300565.1 | NP_001287494.1 | 4 - 5 | 1639 | 92 | ||
NM_001316508.1 | NP_001303437.1 | 4 - 5 | 1639 | 92 | ||
NM_001316509.1 | NP_001303438.1 | 5 - 6 | 1825 | 92 | ||
NM_142949.4 | NP_651206.3 | 5 - 6 | 1825 | 92 | ||
NM_170115.3 | NP_732941.2 | 5 - 6 | 1825 | 92 | ||
GenBank mRNA | AY069522.1 | - | 5 - 6 | 1307 | 92 | |
BT044258.1 | - | 5 - 6 | 1822 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |