Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143066.3 | NP_651323.3 | 1 - 2 | 258 | 61 | |
GenBank mRNA | AY075503.1 | - | 1 - 2 | 257 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |