Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143133.3 | NP_651390.2 | 2 - 3 | 747 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |